missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nkx1.2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15877193
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15877193 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877193 Supplier Invitrogen™ Supplier No. PA568784

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Pig: 86%; Rat: 92%.

NKX1-2 may function in cell specification, particularly in the CNS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx1.2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NKX1-2
Gene Accession No. P42580
Gene Alias bB238F13.2; C10orf121; Homeobox protein SAX-1; NK1 homeobox 2; NK1 transcription factor related, locus 2; NK1 transcription factor related, locus 2 (Drosophila); NK1 transcription factor-related protein 2; Nkx1.1; NKX-1.1; Nkx-1.2; NKX11; NKX-11; NKX1-2; Sax1; spinal cord axial homeobox gene 1
Gene Symbols NKX1-2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse Nkx1.2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20231
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.