missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nkx2.1 Monoclonal Antibody (8G7G3/1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA513961

Product Code. 11384953

  • € 552.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-13961 targets Thyroid Transcription Factor 1 in IF and IHC (P) applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13961 immunogen is rat TTF-1 recombinant protein.

This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nkx2.1
Monoclonal
0.2 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4
P23441, P43699, P50220
NKX2-1
Rat TTF-1 recombinant protein.
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG1 κ
Immunohistochemistry (Paraffin)
8G7G3/1
Unconjugated
NKX2-1
AV026640; BCH; BHC; homeobox protein NK-2 homolog A; Homeobox protein Nkx-2.1; NK-2; NK2 homeobox 1; NK-2 homolog A; NKX2.1; Nkx-2.1; NKX2-1; NKX2A; NMTC1; RNA polymerase I transcription termination factor; T/EBP; TEBP; Thyroid nuclear factor 1; thyroid transcription factor 1; thyroid transcription factor 1 TTF-1 NK-2; thyroid transcription factor 1-like; thyroid-specific enhancer-binding protein; tinman; TITF1; TTF1; TTF-1
Mouse
Protein A/G
RUO
21869, 25628, 7080
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.