missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2-1, Rabbit anti-Human, Clone: G21-G, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB15954.200uL
This item is not returnable.
View return policy
Specifications
| NKX2-1 | |
| Monoclonal | |
| Unconjugated | |
| In 20 mM Tris-HCl buffer, pH 8.0 (20 mg/mL BSA, 0.05% Sodium Azide). | |
| BCH/BHC/NK-2/NKX2.1/NKX2A/TEBP/TITF1/TTF-1/TTF1 | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |
| Immunohistochemistry (Paraffin) | |
| G21-G | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user. | |
| NK2 homeobox 1 | |
| NKX2-1 | |
| A synthetic peptide corresponding to N-terminus of human NKX2-1. | |
| RUO | |
| 7080 | |
| Store at 4°C. Do not freeze. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction