missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.2 Antibody (3E4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37486
This item is not returnable.
View return policy
Description
NKX2.2 Monoclonal specifically detects NKX2.2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| NKX2.2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B | |
| Mouse | |
| 30.1 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 30000 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| NKX2-2 | |
| Purified recombinant fragment of human NKX2.2 expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 4821 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction