Learn More
NKX2.2 Antibody (CL11380), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21191-25ul
 View more versions of this product
Product Code. 18642685
  • € 305.00 / 25µL
Description
NKX2.2 Monoclonal antibody specifically detects NKX2.2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKX2.2
Monoclonal
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
4821
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
CL11380
Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B
This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of O95096. Actual immunogen sequence is proprietary.
25 μg