missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.2 Antibody (rNX2/294), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54345AF350
This item is not returnable.
View return policy
Description
NKX2.2 Monoclonal antibody specifically detects NKX2.2 in Human, Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
| NKX2.2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B | |
| Recombinant full-length human NKX2.2 protein (Uniprot: O95096) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, CyTOF | |
| rNX2/294 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4821 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction