missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nkx2.2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578079
This item is not returnable.
View return policy
Description
Positive Control: Nkx2.2-transfected 293T Predicted Reactivity: Mouse (100%), Chicken (89%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
Specifications
| Nkx2.2 | |
| Polyclonal | |
| Unconjugated | |
| NKX2-2 | |
| Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18088, 366214 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 1.13 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P42586 | |
| NKX2-2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of mouse Nkx2.2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction