Learn More
Invitrogen™ Nkx2.2 Recombinant Rabbit Monoclonal Antibody (24GB715)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557122
 View more versions of this product
Product Code. 30275357
  • € 151.00 / 20µL
Description
Nkx2.2 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene contains a homeobox domain and may be involved in the morphogenesis of the central nervous system. This gene is found on chromosome 20 near NKX2-4, and these two genes appear to be duplicated on chromosome 14 in the form of TITF1 and NKX2-8. The encoded protein is likely to be a nuclear transcription factor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nkx2.2
Recombinant Monoclonal
Unconjugated
NKX2-2
Drosophila NK2 transcription factor related, locus 2; glycoprotein GP330, renal; homeobox protein NK-2 homolog B; Homeobox protein Nkx-2.2; homeobox protein Nkx2-2; NK2 homeobox 2; NK2 transcription factor related locus 2; NK2 transcription factor related, locus 2; NK2 transcription factor-like protein B; NKX2.2; Nkx-2.2; Nkx2\.2; Nkx2-2; NKX2B; tinman
Rabbit
Affinity chromatography
RUO
18088, 4821
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB715
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95096, P42586
NKX2-2
A synthesized peptide derived from human NKX2.2 (aa 1-140).
20 μL
Primary
Human, Mouse