missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nkx2.3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578641
This item is not returnable.
View return policy
Description
Positive Control: human NKX2-3-transfected 293T Predicted Reactivity: Mouse (83%), Rat (84%), Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NKX2-3 encodes a homeodomain-containing transcription factor. The encoded protein is a member of the NKX family of homeodomain transcription factors. Studies of similar proteins in mouse and rat have indicated a potential role in cellular differentiation.
Specifications
| Nkx2.3 | |
| Polyclonal | |
| Unconjugated | |
| NKX2-3 | |
| CSX3; Drosophila NK2 transcription factor related, locus 3; homeobox protein NK-2 homolog 3; Homeobox protein NK-2 homolog C; homeobox protein Nkx-2.3; homeobox protein Nkx-2.3; LOW QUALITY PROTEIN: homeobox protein Nkx-2.3; hypothetical protein LOC539451; NK2 homeobox 3; NK2 transcription factor related, locus 3; NK2.3; NKX2.3; Nkx-2.3; NKX23; NKX2-3; NKX2C; Nkx2-C; NKX4-3; tinman | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 159296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8TAU0 | |
| NKX2-3 | |
| Recombinant protein encompassing a sequence within the center region of human NKX2-3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction