missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.8 Antibody (NKX28/2548), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08638AF750
This item is not returnable.
View return policy
Description
NKX2.8 Monoclonal antibody specifically detects NKX2.8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| NKX2.8 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H | |
| Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary) (Uniprot: O15522) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| NKX28/2548 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 26257 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction