missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2 Monoclonal Antibody (OTI15A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803319S
This item is not returnable.
View return policy
Description
NKX2 Monoclonal antibody specifically detects NKX2 in Human samples. It is validated for Western BlotSpecifications
| NKX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9P0M6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55506 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI15A10 | |
| Unconjugated | |
| H2AFY2 | |
| AV026640; BCH; BHC; homeobox protein NK-2 homolog A; Homeobox protein Nkx-2.1; NK-2; NK2 homeobox 1; NK-2 homolog A; NKX2.1; Nkx-2.1; NKX2-1; NKX2A; NMTC1; RNA polymerase I transcription termination factor; T/EBP; TEBP; Thyroid nuclear factor 1; thyroid transcription factor 1; thyroid transcription factor 1 TTF-1 NK-2; thyroid transcription factor 1-like; thyroid-specific enhancer-binding protein; tinman; TITF1; TTF1; TTF-1 | |
| Human recombinant protein fragment corresponding to amino acids 2-261 of human H2AFY2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction