Learn More
NKX2 Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803245S
Encompass_Preferred
 View more versions of this product
Product Code. 17532203
  • € 242.00 / 30µL
Description
NKX2 Monoclonal antibody specifically detects NKX2 in Human samples. It is validated for Western Blot
Specifications
Specifications
NKX2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P35398
Mouse
Affinity chromatography
RUO
6095
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1G2
Unconjugated
RORA
AV026640; BCH; BHC; homeobox protein NK-2 homolog A; Homeobox protein Nkx-2.1; NK-2; NK2 homeobox 1; NK-2 homolog A; NKX2.1; Nkx-2.1; NKX2-1; NKX2A; NMTC1; RNA polymerase I transcription termination factor; T/EBP; TEBP; Thyroid nuclear factor 1; thyroid transcription factor 1; thyroid transcription factor 1 TTF-1 NK-2; thyroid transcription factor 1-like; thyroid-specific enhancer-binding protein; tinman; TITF1; TTF1; TTF-1
Human recombinant protein fragment corresponding to amino acids 98-345 of human RORA produced in E.coli.
30 μL