missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2 Monoclonal Antibody (OTI15A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803319
This item is not returnable.
View return policy
Description
This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| NKX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P43699 | |
| NKX2-1 | |
| Full length human recombit protein of human NKX2-1 ( NP_003308) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI15A10 | |
| Unconjugated | |
| NKX2-1 | |
| BCH, BHC, NK-2, NKX2.1, NKX2A, NMTC1, T/EBP, TEBP, TITF1, TTF-1, TTF1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7080 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction