missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX3.1 Antibody (0361), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-1828AF750
This item is not returnable.
View return policy
Description
NKX3.1 Monoclonal specifically detects NKX3.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| NKX3.1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NKX3-1 | |
| Full-length human Nkx3.1 produced in bacteria as a hexahistidine-tagged protein. [UniProtNo. Q99801] | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| KnockDown, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 361 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A | |
| Mouse | |
| 0.1 mL | |
| 4824 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction