missing translation for 'onlineSavingsMsg'
Learn More

NFIA Monoclonal Antibody (OTI4G7), TrueMAB™, OriGene

Product Code. 17524994
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17524994 30 μL 30µL
17504994 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17524994 Supplier Origene Technologies Supplier No. TA811198S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NFIA Monoclonal antibody specifically detects NFIA in Human samples. It is validated for Western Blot

Specifications

Antigen NFIA
Applications Western Blot
Classification Monoclonal
Clone OTI4G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ELF1
Gene Accession No. P32519
Gene Alias 1110047K16Rik; 9430022M17Rik; CCAAT-box-binding transcription factor; CTF; KIAA1439; NF1A; NF1-A; Nf1l21; NF-I/A; Nfia; NFI-A; NFI-L; Nuclear factor 1 A-type; nuclear factor 1/A; nuclear factor I A; nuclear factor I/A; TGGCA-binding protein; Unknown (protein for MGC:128467)
Host Species Mouse
Immunogen Full length human recombinant protein of human ELF1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1997
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.