missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX3.1 Mouse anti-Human, Clone: NKX3.1/3347, Novus Biologicals™
Mouse Monoclonal Antibody
€ 484.00
Specifications
| Antigen | NKX3.1 |
|---|---|
| Clone | NKX3.1/3347 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | ELISA, Immunohistochemistry (Paraffin), Peptide Array |
Description
NKX3.1 Monoclonal specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.Specifications
| NKX3.1 | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 4824 | |
| IgG2b κ | |
| Protein A or G purified |
| NKX3.1/3347 | |
| ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A | |
| Recombinantfragment(around aa 92-224) of humanNKX3.1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title