Learn More
Nkx3.1 Mouse anti-Human, Clone: OTI9H7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF805433
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.Specifications
| Nkx3.1 | |
| Monoclonal | |
| Unconjugated | |
| NKX3-1 | |
| BAPX2, NKX3, NKX3.1, NKX3A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4824 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9H7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q99801 | |
| NKX3-1 | |
| Full length human recombit protein of human NKX3-1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.