missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nkx3.1 Recombinant Rabbit Monoclonal Antibody (RM430)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544589
This item is not returnable.
View return policy
Description
This antibody reacts to human EGFR (L858R).
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
Specifications
| Nkx3.1 | |
| Recombinant Monoclonal | |
| 400 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Q99801 | |
| NKX3-1 | |
| A peptide corresponding to the N-terminus of human Nkx3.1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| RM430 | |
| Unconjugated | |
| NKX3-1 | |
| bagpipe; BAPX2; Bax; Drosophila NK-3 transcription factor, locus 1; homeobox protein NK-3 homolog A; Homeobox protein Nkx-3.1; NK homeobox, family 3, A; NK3 homeobox 1; NK3 transcription factor homolog A; NK3 transcription factor related, locus 1; NK-3 transcription factor, locus 1; NK-3 transcription factor, locus 1 (Drosophila); NKX3; NKX3.1; Nkx-3.1; Nkx3-1; NKX3-1 antibody; NKX3A; prostate-specific ag | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4824 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction