missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX3-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 189.00
Specifications
| Antigen | NKX3-2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
NKX3-2 Polyclonal specifically detects NKX3-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NKX3-2 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_001180 | |
| 579 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the center of human BAPX1. The immunogen is located within amino acids 180 - 230 of BAPX1. Amino Acid Squence: EEEPAAPKPRKKRSRAAFS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Bagpipe homeobox protein homolog 1, BAPX1bagpipe homeobox homolog 1 (Drosophila), Homeobox protein NK-3 homolog B, homeobox protein Nkx-3.2, MGC138171, NK3 homeobox 2, NKX3.2, NKX3BSMMD | |
| NKX3-2 | |
| IgG | |
| Affinity Purified | |
| BAPX1 antibody is predicted not to cross-react with other NKX homeobox proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title