missing translation for 'onlineSavingsMsg'
Learn More

NKX6.1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97468MFV500

Product Code. 18528383

  • € 451.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NKX6.1 Polyclonal antibody specifically detects NKX6.1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NKX6.1
Polyclonal
Immunocytochemistry/ Immunofluorescence
Homeobox protein NK-6 homolog A, NK homeo box, family 6, member A, NK homeobox (Drosophila), family 6, A, NK homeobox, family 6, A, NK6 homeobox 1, NK6 transcription factor homolog A, NK6 transcription factor related, locus 1, NK6 transcription factor related, locus 1 (Drosophila), NKX6.1, NKX6Ahomeobox protein Nkx-6.1
Produced in rabbits immunized with E. coli-derived Human NKX6.1 fragment.
0.1 mL
Pancreatic Stem Cell Markers, Stem Cell Signaling Pathway, Transcription Factors and Regulators
4825
Store at 4C in the dark.
IgG
Immunocytochemistry
mFluor Violet 500 SE
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.