missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NLE1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15791781 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15791781 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15791781 Supplier Invitrogen™ Supplier No. PA553850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TIKVWRAHDG VLCRTLQGHG HWVNTMALST DYALRTGAFE PAEASVNPQD LQGSLQELKE RALSRYNLVR GQGPERLVSG SDDFTL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

NLE1 plays a role in regulating Notch activity, in regulating the expression of CDKN1A and several members of the Wnt pathway, probably via its effects on Notch activity. NLE1 is required during embryogenesis for inner mass cell survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLE1
Gene Accession No. Q9NVX2
Gene Alias AL022765; BC018399; FLJ10458; HUSSY-07; l11Jus1; l11Jus4; Nle; Nle1; Notchless gene homolog; notchless homolog 1; notchless homolog 1 (Drosophila); notchless protein homolog 1; novel WD40 repeat containing protein; RGD1308301
Gene Symbols NLE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NLE1. Recombinant protein control fragment (Product #RP-92396).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54475
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.