Learn More
NLGN1 Mouse anti-Human, Unconjugated, Clone: 2D1A12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66964-1-IG-150UL
 View more versions of this product
Product Code. 16859295
  • € 410.00 / 150µL
Description
The NLGN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NLGN1. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The NLGN1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
NLGN1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig, Rabbit
Western Blot, Immunohistochemistry
2D1A12
Unconjugated
150 μL
22871