Learn More
NLGN1 Mouse anti-Human, Unconjugated, Clone: 2D1A12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66964-1-IG-20UL
Description
The NLGN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NLGN1. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The NLGN1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| NLGN1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit |
| Western Blot, Immunohistochemistry | |
| 2D1A12 | |
| Unconjugated | |
| 20 μL | |
| 22871 | |
| IgG1 |