Learn More
Invitrogen™ NLGN3 Monoclonal Antibody (N110/29), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545697
Description
Mouse: 99% identity (118/119 amino acids identical). Human: 98% identity (116/119 amino acids identical) ~60% identity with Neuroligin-1. ~40% identity with Neuroligin-2. 1 μg/mL of MA5-45697 was sufficient for detection of Neuroligin 3 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 110kDa. Does not cross-react with Neuroligin-1, -2, -4 or -4. This antibody was formerly sold as clone S110-29.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| NLGN3 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q62889, Q8BYM5, Q9NZ94 | |
| NLGN3 | |
| Fusion protein amino acids 730-848 (intracellular C-terminus) of rat Neuroligin-3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N110/29 | |
| APC | |
| NLGN3 | |
| A230085M13Rik; ASPGX1; AUTSX1; Gliotactin homolog; HNL3; KIAA1480; neuroligin 3; neuroligin 3 protein; neuroligin-3; NL3; NLGN3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 171297, 245537, 54413 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.