missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nlgn4l, Mouse anti-Mouse, ATTO594, Clone: S98-7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB18670.100ug
This item is not returnable.
View return policy
Specifications
| Nlgn4l | |
| Monoclonal | |
| Atto 594 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| NL-4/NL4/Nlgn4/Nlgn4x | |
| Mouse | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S98-7 | |
| Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| neuroligin 4-like | |
| Nlgn4l | |
| Recombinant protein corresponding to amino acids 782-945 at C-terminus of mouse Nlgn4l. | |
| RUO | |
| 100113365 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction