missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99830-100ul
This item is not returnable.
View return policy
Description
NLK Polyclonal antibody specifically detects NLK in Human samples. It is validated for Western Blot, ELISA
Specifications
| NLK | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| DKFZp761G1211, EC 2.7.11, EC 2.7.11.24, FLJ21033, LAK1, nemo like kinase, nemo-like kinaseserine/threonine-protein kinase NLK, Protein LAK1 | |
| Produced in rabbits immunized with purified, recombinant Human NLK (Uniprot#: Q9UBE8; Val121-Glu527) | |
| 100 μg | |
| Protein Kinase, Wnt Signaling Pathway | |
| 51701 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction