Learn More
Eurofins DiscoverX NLK Protein, Active
Description
NLK (Nemo Like Kinase) regulates a number of transcription factors with key roles in cell fate determination. It enables ubiquitin protein ligase binding activity and Iinvolved in protein stabilization and transforming growth factor beta receptor signaling pathway. It predicted to be located in cytosol and nucleoplasm, and to be active in cytoplasm and nucleus.
GenBank NM_016231
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 48.7 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥53% |
| Protein | NLK, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.