Learn More
Invitrogen™ NLN Monoclonal Antibody (OTI3H4)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525881
Product Code. 15915484
  • € 572.00 / 100µL
Description
NLN Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NLN
Monoclonal
0.92 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P42676, Q91YP2, Q9BYT8
Nln
Full length human recombinant protein of NLN produced in HEK293T cell.
100 μL
Primary
Human, Mouse, Rat, Canine, Monkey
Antibody
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3H4
Unconjugated
Nln
4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN
Mouse
Affinity Chromatography
RUO
117041, 478081, 57486, 75805
-20°C, Avoid Freeze/Thaw Cycles