Learn More
NLN Mouse anti-Human, Rat, Clone: OTI3G8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504402
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Neurolysin (EC 3.4.24.16) is an enzyme involved in the metabolic inactivation of bioactive peptides.Specifications
| NLN | |
| Monoclonal | |
| Unconjugated | |
| NLN | |
| AGTBP, EP24.16, MEP, MOP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 117041, 57486 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3G8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P42676, Q9BYT8 | |
| NLN | |
| Full length human recombit protein of human NLN produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.