Learn More
Invitrogen™ NLN Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA548037
Description
This antibody detects human, mouse, and rat Neurolysin Neurolysin in Western blots and human Neurolysin in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Meprin alpha, rhMeprin beta subunit, and rhTHOP1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
Specifications
| NLN | |
| Polyclonal | |
| Unconjugated | |
| Nln | |
| 4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 117041, 57486, 75805 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P42676, Q91YP2, Q9BYT8 | |
| Nln | |
| E. coli-derived recombinant human Neurolysin (aa 38-704). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.