missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 149.35 - € 422.30
Specifications
| Antigen | NLN |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Neurolysin (EC 3.4.24.16) is an enzyme involved in the metabolic inactivation of bioactive peptides.Specifications
| NLN | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P42676, Q91YP2, Q9BYT8 | |
| 117041, 57486, 75805 | |
| NLN Fusion Protein Ag6524 | |
| Primary | |
| -20°C | |
| NLN |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AGTBP, Angiotensin binding protein, KIAA1226, MEP, Microsomal endopeptidase, Mitochondrial oligopeptidase M, Neurolysin, mitochondrial, Neurotensin endopeptidase, NLN | |
| NLN | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title