missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRC3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41123JF549
This item is not returnable.
View return policy
Description
NLRC3 Polyclonal antibody specifically detects NLRC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| NLRC3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Janelia Fluor 549 | |
| CARD15-Like Protein, Caterpiller Protein 16.2, CLR16.2, Leucine Rich Repeat And CARD Domain Containing 3, NLR Family, CARD Domain Containing 3, NOD3, NOD-Like Receptor C3, Nucleotide-Binding Oligomerization Domain Protein 3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And CARD Domain Containing 3, Protein NLRC3 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human NOD3. The immunogen is located within amino acids 630 - 680 of NOD3. Amino Acid Squence: CRKLRLDTNQFQDPVMEL | |
| 0.1 mL | |
| Innate Immunity | |
| 197358 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction