missing translation for 'onlineSavingsMsg'
Learn More

Nlrc4 (phospho S533) Rat anti-Human, Mouse, Clone:4B7B7, Abnova™

Product Code. 16064616
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16064616 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16064616 Supplier Abnova Supplier No. MAB17177.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat monoclonal antibody raised against synthetic peptide of mouse Nlrc4 (phospho S533).

Specifications

Antigen Nlrc4 (phospho S533)
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4B7B7
Conjugate Unconjugated
Description NLR family, CARD domain containing 4
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry (1:200-1:1000)
Flow Cytometry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene Nlrc4
Gene Alias 9530011P19Rik, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, Card12, IPAF
Host Species Rat
Immunogen A synthetic peptide corresponding to amino acid 525-538 of human Nlrc4 (phospho S533).
Quantity 100 μg
Gene ID (Entrez) 268973
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.