missing translation for 'onlineSavingsMsg'
Learn More

NLRP1/NALP1 Antibody, FITC, Novus Biologicals™

Product Code. 18544283 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18544283 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18544283 Supplier Novus Biologicals Supplier No. NBP176656F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NLRP1/NALP1 Polyclonal antibody specifically detects NLRP1/NALP1 in Human samples. It is validated for ELISA, Immunofluorescence, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP1/NALP1
Applications ELISA, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias CARD7SLEV1, Caspase recruitment domain-containing protein 7, CLR17.1, Death effector filament-forming ced-4-like apoptosis protein, DEFCAPVAMAS1, DKFZp586O1822, KIAA0926DEFCAP-L/S, NACHT, leucine rich repeat and PYD (pyrin domain) containing 1, NACHT, leucine rich repeat and PYD containing 1, NACHT, LRR and PYD containing protein 1, NACHT, LRR and PYD domains-containing protein 1, NACPP1044, NALP1caspase recruitment domain protein 7, NLR family, pyrin domain containing 1, Nucleotide-binding domain and caspase recruitment domain, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 1
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the carboxy terminus of human NALP1. The immunogen is located within the last 50 amino acids of NALP1. Amino Acid Squence: EKGSKKGLLPLSS
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 22861
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.