missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP13 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76294H
This item is not returnable.
View return policy
Description
NLRP13 Polyclonal antibody specifically detects NLRP13 in Human samples. It is validated for Western Blot, ELISA
Specifications
| NLRP13 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| CLR19.7, leucine rich repeat and pyrin domain containing 13, NACHT, leucine rich repeat and PYD containing 13, NACHT, LRR and PYD containing protein 13, NACHT, LRR and PYD domains-containing protein 13, NALP13, NLR family, pyrin domain containing 13, NOD14, nucleotide-binding oligomerization domain protein 14, PAN13 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human NALP13. The immunogen is located within amino acids 150 - 200 of NALP13. Amino Acid Squence: QDPNQEEPEMLEEADHRR | |
| 0.1 mL | |
| Innate Immunity | |
| 126204 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction