missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP3, Mouse anti-Human, Clone: Nalpy3-b, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16140.50ug
This item is not returnable.
View return policy
Specifications
| NLRP3 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (0.02% sodium azide). | |
| AGTAVPRL/AII/AII/AVP/AVP/C1orf7/CIAS1/CLR1.1/FCAS/FCU/FLJ95925/MWS/NALP3/PYPAF1 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Nalpy3-b | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunohistochemistry (Frozen sections) Immunoprecipitation (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| NLR family, pyrin domain containing 3 | |
| NLRP3 | |
| Recombinant protein corresponding to human NLRP3. | |
| RUO | |
| 114548 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction