missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NLRP6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30535579
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30535579 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535579 Supplier Invitrogen™ Supplier No. PA5149510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NLRP6 Polyclonal Antibody for Western Blot, IHC (P)

NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP6, also known as PYPAF5, is highly expressed in granulocytes and T cells and co-localizes with the PYRIN-CARD protein ASC. Co-expression of NALP6 with ASC results in a synergistic activation of NF-kappa-B and activated caspase-1-dependent cytokine processing, suggesting that NALP6 functions in immune cells to coordinate the transduction of pro-inflammatory signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene NLRP6
Gene Accession No. P59044
Gene Alias AI504961; angiotensin II/vasopressin receptor; angiotensin/vasopressin receptor; AVR; CLR11.4; dual angiotensin II/vasopressin receptor; NACHT, leucine rich repeat and PYD containing 6; NACHT, LRR and PYD domains-containing protein 6; NACHT/LRR/pyrin domain-containing protein 6; NALP6; Navr; Navr/Avr; NLR family pyrin domain containing 6; NLR family, pyrin domain containing 6; Nlrp6; Non-angiotensin-vasopressin receptor; Non-AVR; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 6; PAN3; Pypaf5; PYRIN-containing APAF1-like protein 5; PYRIN-containing APAF1-like protein 5-like
Gene Symbols NLRP6
Host Species Rabbit
Immunogen Partial recombinant protein made to an N-terminal portion of human NALP6 (between amino acids 10-200).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171389
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.