Learn More
NLRP7 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27897-1-AP-20UL
This item is not returnable.
View return policy
Description
The NLRP7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NLRP7. This antibody recognizes human antigen. The NLRP7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| NLRP7 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| CLR19.4, HYDM, NALP7, NLRP7, NOD12, PAN7, PYPAF3 | |
| 20 μL | |
| 199713 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |