Learn More
NLRP7 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27897-1-AP-20UL
 View more versions of this product
Product Code. 18066405
  • € 145.00 / 20µL
Description
The NLRP7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NLRP7. This antibody recognizes human antigen. The NLRP7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
NLRP7
400 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
CLR19.4, HYDM, NALP7, NLRP7, NOD12, PAN7, PYPAF3
20 μL
199713
Store at -20°C. Stable for one year after shipment.