Learn More
Schmidlin™ NM Plus FID Station H2 and Zero Air Generator
Click to view available options
Flow Rate:
100 mL/min.
1000 mL/min.
300 mL/min.
Wattage:
90 W
150 W
400 W
Unit Size:
Each
Description
- Increase efficiency and improve performance
- On-demand supply 24/7
- Zero air <0.1 ppm
- THC Pressure: 11 barg (160 psig)
- Permeation membrane drying system
- USB connectivity
- Easy to install with complete 'plug and play' functionality
- Ideal for all GC and GC/MS applications
Specifications
Specifications
| Depth (Metric) | 68 cm |
| Flow Rate | 1000 mL/min. |
| Height (Metric) | 15 cm |
| Max. Ambient Operating Temperature | 35°C |
| Outlet Pressure | 11 bar |
| Width (English) | 27 in. |
| Width (Metric) | 69 cm |
| Connections | Outlet: 1/8 in. |
| Electrical Requirements | 110/120 V at 60 Hz, 220/240 V at 50 Hz |
| Nitrogen Purity | >99.99996% |
| Show More |