missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM23-H1 Antibody (084), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89930AF700
This item is not returnable.
View return policy
Description
NM23-H1 Monoclonal specifically detects NM23-H1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NM23-H1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human NM23-H1 (NP_000260.1; Ala2-Glu152). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 4830 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 84 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction