missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM23-H1 Antibody (5F4), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59498JF646
This item is not returnable.
View return policy
Description
NM23-H1 Monoclonal antibody specifically detects NM23-H1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| NM23-H1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein | |
| Recombinant human NM23-H1 (1-152aa) purified from E. coli | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 4830 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 5F4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction