missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NM23-H2/NME2 Antibody (NME2/6434) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26978JF646
This item is not returnable.
View return policy
Description
NM23-H2/NME2 Monoclonal antibody specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Specifications
| NM23-H2/NME2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
| Recombinant full-length human NM23-H2/NME2 protein | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 4831 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| NME2/6434 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction