missing translation for 'onlineSavingsMsg'
Learn More

NM23-H2/NME2 Antibody (NME2/6435), Novus Biologicals™

Mouse Monoclonal Antibody

€ 249.00 - € 494.00

Specifications

Antigen NM23-H2/NME2
Clone NME2/6435
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18621695
View Documents
Novus Biologicals
NBP3-20542-100ug
100 μg
€ 494.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18609584
View Documents
Novus Biologicals
NBP3-20542-20ug
20 μg
€ 249.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NM23-H2/NME2 Monoclonal antibody specifically detects NM23-H2/NME2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

NM23-H2/NME2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
10mM PBS with 0.05% BSA
4831
IgG1 κ
Protein A or G purified
NME2/6435
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in
Recombinant full-length human NM23-H2/NME2 protein
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.