missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDA R, NR1 Subunit Antibody (1031915), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB10655B
This item is not returnable.
View return policy
Description
NMDA R, NR1 Subunit Monoclonal antibody specifically detects NMDA R, NR1 Subunit in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| NMDA R, NR1 Subunit | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 1031915 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| GluN1, MRD8, NDHMSD, NDHMSR,NMD-R1, NMDA1, NMDAR1, NR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GRIN1/NMDAR1, Met1-Gln559 | |
| 0.1 mL | |
| Cellular Signaling, Neuroscience | |
| 2902 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction