missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMDAR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585751
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%). Positive Control: mouse brain, mouse fetal brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.
Specifications
| NMDAR1 | |
| Polyclonal | |
| Unconjugated | |
| GRIN1 | |
| GluN1; Glun1r; GluRdelta1; Glurz1; GluRzeta1; glutamate [NMDA] receptor subunit zeta 1; glutamate [NMDA] receptor subunit zeta-1; glutamate ionotropic receptor NMDA type subunit 1; glutamate receptor; glutamate receptor ionotropic, NMDA 1; glutamate receptor subunit zeta 1; glutamate receptor, ionotropic, NMDA1 (zeta 1); glutamate receptor, ionotropic, N-methyl D-aspartate 1; GRIN1; M100174; MRD8; neurotransmitter receptor; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDA1; Nmdar; NMDAR1; NMD-R1; NMDZ1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor 1; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; NMZ1; NR1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; Rgsc174; RP11-350O14.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14810, 24408, 494007 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.37 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P35438, P35439, Q5R1P0 | |
| GRIN1 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of mouse NMDAR1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat, Canine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction