missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2A/GRIN2A Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | NMDAR2A/GRIN2A |
|---|---|
| Concentration | 800 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235295
|
Proteintech
28571-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30238139
|
Proteintech
28571-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NMDAR2A/GRIN2A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NMDAR2A/GRIN2A. This antibody recognizes mouse, rat antigen. The NMDAR2A/GRIN2A antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| NMDAR2A/GRIN2A | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| NMDAR2A; NR2A | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 800 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 2903 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title