missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2A, p Tyr1325 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29535
This item is not returnable.
View return policy
Description
NMDAR2A Polyclonal specifically detects NMDAR2A in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NMDAR2A (p Tyr1325) | |
| Polyclonal | |
| Western Blot 1:1000 | |
| Q00959 | |
| GRIN2A | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr1325 conjugated to KLH. Accession # Q00959 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼180k NMDAR NR2A-subunit protein phosphorylated at Tyr1325 in Western blots. Immunolabeling is completely blocked by blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| GluN2A, glutamate receptor, ionotropic, N-methyl D-aspartate 2A, hNR2A, NMDA receptor subtype 2A, N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor subunit 2A, subunit epsilon-1 | |
| Rabbit | |
| 180 kDa | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 2903 | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction