missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMDAR2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143633
This item is not returnable.
View return policy
Description
Specific for endogenous levels of the ~180 kDa NR2B subunit of the NMDA receptor. Immunolabeling is blocked by preadsorption of antibody with the fusion protein used to generate the antibody. No reactivity towards the NR2A and NR2C subunits.
NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer's Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal dominant 6-related mental retardation.
Specifications
| NMDAR2B | |
| Polyclonal | |
| Unconjugated | |
| GRIN2B | |
| AW490526; EIEE27; GluN2B; GluRepsilon2; glutamate [NMDA] receptor subunit epsilon-2; glutamate ionotropic receptor NMDA type subunit 2B; glutamate receptor; glutamate receptor ionotropic, NMDA 2B; glutamate receptor subunit epsilon-2; glutamate receptor, ionotropic, NMDA2B; glutamate receptor, ionotropic, NMDA2B (epsilon 2); glutamate receptor, ionotropic, N-methyl D-aspartate 2B; GRIN 2B; GRIN2B; HGNC:4586; hNR 3; hNR3; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2B; MGC142178; MGC142180; MRD6; Nmdar2b; NMDE2; NME2; N-methyl D-aspartate receptor 2B; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 2B; N-methyl-D-aspartate receptor subunit 3; NR2B; NR3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14812, 24410, 2904 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| 10mM HEPES with 100μg/mL BSA, 50% glycerol, 150mM NaCl and no preservative; pH 7.5 | |
| Q00960, Q01097, Q13224 | |
| GRIN2B | |
| Fusion protein from the C-terminal region of the NR2B subunit of the rat NMDA receptor. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction