missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMDAR2C Polyclonal Antibody

Description
Antibody detects endogenous levels of total NMDA epsilon 3.
The NMDA Receptor (NMDAR) plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimers, epilepsy and ischemic neuronal cell death. The NMDA receptor is also one of the principal molecular targets for alcohol in the CNS. The NMDAR is also potentiated by protein phosphorylation. The NR1 protein can form NMDA activated channels when expressed in Xenopus oocytes but the currents in such channels are much smaller than those seen in situ. Channels with more physiological characteristics are produced when the NR1 subunit is combined with one or more of th NMDAR2 (NR2 A-D) subunits.
Specifications
Specifications
| Antigen | NMDAR2C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GRIN2C |
| Gene Accession No. | Q14957 |
| Gene Alias | GluN2C; GluRepsilon3; glutamate [NMDA] receptor subunit epsilon-3; glutamate ionotropic receptor NMDA type subunit 2C; glutamate receptor; glutamate receptor ionotropic, NMDA 2C; glutamate receptor ionotropic, NMDA 2C; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2C; glutamate receptor, ionotropic, NMDA2C; glutamate receptor, ionotropic, NMDA2C (epsilon 3); glutamate receptor, ionotropic, N-methyl D-aspartate 2C; Grin2c; LOW QUALITY PROTEIN: glutamate receptor ionotropic, NMDA 2C; NMDA glutamate receptor; NMDAR2C; N-methyl D-aspartate receptor subtype 2C; N-methyl-D-aspartate receptor subunit 2C; NR2; NR2C |
| Gene Symbols | GRIN2C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?