missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMDAR2D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101608
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GRIN2D.
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localized with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate, whereas the NMDA receptors exhibit slow kinetsis of Ca2+ ions and a high permeability for Ca2+ ions. The NMDA receptors consist of five subunits: epsilon 1, 2, 3, 4 and one zeta subunit. The zeta subunit is expressed throughout the brainstem whereas the four epsilon subunits display limited distribution.
Specifications
| NMDAR2D | |
| Polyclonal | |
| Unconjugated | |
| GRIN2D | |
| EB11; estrogen receptor binding CpG island; GluN2D; glur epsilon 4; GluRepsilon4; glutamate [NMDA] receptor subunit epsilon-4; glutamate ionotropic receptor NMDA type subunit 2D; glutamate receptor; glutamate receptor ionotropic, NMDA 2D; glutamate receptor, ionotropic, NMDA2D; glutamate receptor, ionotropic, NMDA2D (epsilon 4); glutamate receptor, ionotropic, N-methyl D-aspartate 2D; GRIN2D; ionotropic NMDA glutamate receptor epsilon 4 subunit type 2; NMDAR2D; N-methyl D-aspartate receptor subtype 2D; N-methyl-d-aspartate receptor subunit 2D; NR2D | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14814, 24412, 2906 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15399, Q03391, Q62645 | |
| GRIN2D | |
| A synthesized peptide derived from human GRIN2D(Accession O15399), corresponding to amino acid residues Y673-R723. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction